siRNA Details
| Gene Symbol: Wfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023395.2 | NP_075884.1 | 3, 4 | 566 | ||
| GenBank mRNA | AK009971.1 | - | 3, 4 | 569 | ||
| AK018575.1 | - | 3, 4 | 427 | |||
| AK132541.1 | - | 3, 4 | 567 | |||
| AK142707.1 | - | 3, 4 | 435 | |||
| AK161606.1 | - | 3, 4 | 432 | |||
| BC038122.1 | - | 3, 4 | 408 | |||
| BC132073.1 | - | 3, 4 | 495 | 40130350 | ||
| BC132469.1 | - | 3, 4 | 495 | 40130746 | ||

Wfdc1
Genomic Map