siRNA Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026430.3 | NP_080706.1 | 10 | 796 | ||
| GenBank mRNA | AF399958.1 | - | 10 | 796 | ||
| AK005536.1 | - | 10 | 736 | |||
| AK075806.1 | - | 10 | 848 | |||
| AK076166.1 | - | 9 | 837 | |||
| AK152376.1 | - | 10 | 851 | |||
| BC037049.1 | - | 10 | 842 | 4935237 | ||

Uxs1
Genomic Map