siRNA Details
| Gene Symbol: Camsap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081360.1 | NP_001074829.1 | 3 | 1086 | ||
| GenBank mRNA | AK028542.1 | - | 3 | 1087 | ||
| AK053092.1 | - | 3 | 1086 | |||
| BC138367.1 | - | 3 | 680 | 8861387 | ||
| BC145258.1 | - | 3 | 680 | 9053811 | ||

Camsap2
Genomic Map