siRNA Details
| Gene Symbol: Saraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026432.3 | NP_080708.3 | 3 | 840 | ||
| GenBank mRNA | AK007787.1 | - | 3 | 767 | ||
| AK075744.1 | - | 3 | 731 | |||
| AK077118.1 | - | 3 | 827 | |||
| AK088728.1 | - | 3 | 723 | |||
| AK093942.1 | - | 3 | 815 | |||
| AK163218.1 | - | 3 | 773 | |||
| AK206196.1 | - | 11 | ||||
| BC013497.1 | - | 3 | 781 | 4222385 | ||
| BC022616.1 | - | 3 | 818 | 4187981 | ||
| BC024888.1 | - | 3 | 776 | 3988822 | ||

Saraf
Genomic Map