siRNA Details
| Gene Symbol: Saraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026432.3 | NP_080708.3 | 1, 2 | 272 | ||
| GenBank mRNA | AK007787.1 | - | 1, 2 | 199 | ||
| AK075744.1 | - | 1, 2 | 163 | |||
| AK077118.1 | - | 1, 2 | 260 | |||
| AK088728.1 | - | 1, 2 | 155 | |||
| AK093942.1 | - | 1, 2 | 247 | |||
| AK163218.1 | - | 1, 2 | 205 | |||
| BC013497.1 | - | 1, 2 | 213 | 4222385 | ||
| BC022616.1 | - | 1, 2 | 250 | 4187981 | ||
| BC024888.1 | - | 1, 2 | 208 | 3988822 | ||
| BY135425.1 | - | 1, 2 | 272 | |||

Saraf
Genomic Map