siRNA Details
| Gene Symbol: Saraf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026432.3 | NP_080708.3 | 3 | 569 | ||
| GenBank mRNA | AK007787.1 | - | 3 | 496 | ||
| AK075744.1 | - | 3 | 460 | |||
| AK077118.1 | - | 3 | 556 | |||
| AK088728.1 | - | 3 | 452 | |||
| AK093942.1 | - | 3 | 544 | |||
| AK163218.1 | - | 3 | 502 | |||
| AK211246.1 | - | 2 | 150 | |||
| BC013497.1 | - | 3 | 510 | 4222385 | ||
| BC022616.1 | - | 3 | 547 | 4187981 | ||
| BC024888.1 | - | 3 | 505 | 3988822 | ||

Saraf
Genomic Map