siRNA Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026435.5 | NP_080711.1 | 4 | 205 | ||
| GenBank mRNA | AK007788.1 | - | 4 | 169 | ||
| AK008280.1 | - | 4 | 168 | |||
| AK011737.1 | - | 4 | 186 | |||
| AK089948.1 | - | 4 | 171 | |||
| AK148146.1 | - | 4 | 175 | |||
| AK166332.1 | - | 4 | 199 | |||
| AK167301.1 | - | 4 | 174 | |||
| AK167321.1 | - | 4 | 169 | |||
| BC051554.1 | - | 3 | 169 | |||
| BC061065.1 | - | 4 | 204 | 6594260 | ||

Ufm1
Genomic Map