siRNA Details
| Gene Symbol: Ccdc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026439.2 | NP_080715.2 | 2 | 997 | ||
| GenBank mRNA | AB075019.1 | - | 2 | 886 | ||
| AK032359.1 | - | 2 | 968 | |||
| AK132178.1 | - | 2 | 997 | |||
| AK144183.1 | - | 3 | 1221 | |||
| BC022704.1 | - | 1 | 405 | |||
| BC058751.1 | - | 2 | 756 | |||
| BC096487.1 | - | 3 | 1187 | |||

Ccdc80
Genomic Map