siRNA Details
| Gene Symbol: Ccdc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026439.2 | NP_080715.2 | 5 | 2591 | ||
| GenBank mRNA | AB075019.1 | - | 5 | 2480 | ||
| AK009795.1 | - | 2 | 107 | |||
| AK011256.1 | - | 4 | 881 | |||
| AK032359.1 | - | 5 | 2562 | |||
| AK132178.1 | - | 5 | 2591 | |||
| AK144183.1 | - | 6 | 2815 | |||

Ccdc80
Genomic Map