siRNA Details
| Gene Symbol: Pef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026441.4 | NP_080717.2 | 4 | 609 | ||
| GenBank mRNA | AK007394.1 | - | 4 | 551 | ||
| AK013238.1 | - | 4 | 551 | |||
| AK046247.1 | - | 4 | 551 | |||
| AK075978.1 | - | 4 | 614 | |||
| BC019191.1 | - | 4 | 557 | 5029503 | ||

Pef1
Genomic Map