siRNA Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026445.2 | NP_080721.1 | 7, 8 | 701 | ||
| GenBank mRNA | AK013017.1 | - | 7, 8 | 681 | ||
| AK076022.1 | - | 7, 8 | 707 | |||
| AK138300.1 | - | 7, 8 | 692 | |||
| AK140169.1 | - | 8, 9 | 716 | |||
| AK157784.1 | - | 7, 8 | 702 | |||
| AK166216.1 | - | 7, 8 | 680 | |||
| BC119480.2 | - | 5, 6 | 810 | 40050522 | ||

Sumf2
Genomic Map