siRNA Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026447.4 | NP_080723.3 | 9 | 1257 | ||
| NM_198931.3 | NP_945149.2 | 9 | 1261 | |||
| GenBank mRNA | AB474373.1 | - | 9 | 1296 | ||
| AK013149.1 | - | 9 | 1190 | |||
| AK017245.1 | - | 6 | 594 | |||
| AK046387.1 | - | 1156 | ||||
| AK087999.1 | - | 9 | 1295 | |||
| AY332616.1 | - | 9 | 1264 | |||
| BC058248.1 | - | 9 | 1183 | |||

Ppm1m
Genomic Map