siRNA Details
| Gene Symbol: Zfp169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164575.1 | NP_001158047.1 | 5 | 2058 | ||
| NM_026450.3 | NP_080726.3 | 5 | 2090 | |||
| GenBank mRNA | AK006349.1 | - | 1 | 1153 | ||
| AK016063.1 | - | 1 | 1129 | |||
| BC060222.1 | - | 5 | 2048 | 30356624 | ||

Zfp169
Genomic Map