siRNA Details
| Gene Symbol: Spert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164139.1 | NP_001157611.1 | 3 | 439 | ||
| NM_001164140.1 | NP_001157612.1 | 2 | 358 | |||
| NM_001164141.1 | NP_001157613.1 | 3 | 447 | |||
| NM_026457.2 | NP_080733.2 | 2 | 297 | |||
| GenBank mRNA | AK005703.1 | - | 2 | 296 | ||
| AK006329.1 | - | 1 | 27 | |||
| AK007022.1 | - | 2 | 276 | |||
| AY092416.1 | - | 2 | 431 | |||
| BC109147.2 | - | 2 | 276 | 40045644 | ||

Spert
Genomic Map