siRNA Details
| Gene Symbol: MCM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006739.3 | NP_006730.2 | 16, 17 | 2190 | ||
| GenBank mRNA | AK122853.1 | - | 15, 16 | 2038 | ||
| AK130620.1 | - | 15, 16 | 2144 | |||
| AK223323.1 | - | 16, 17 | 2180 | |||
| AK291136.1 | - | 16, 17 | 2171 | |||
| AK298624.1 | - | 16, 17 | 2067 | |||
| AK301121.1 | - | 1658 | ||||
| AK312533.1 | - | 16, 17 | 2170 | |||
| BC000142.1 | - | 16, 17 | 2142 | 2900229 | ||
| BC003656.2 | - | 15, 16 | 2116 | 3542201 | ||
| CR456517.1 | - | 16, 17 | 2130 | |||
| D83986.1 | - | 16, 17 | 2175 | |||
| X74795.2 | - | 15, 16 | 2118 | |||

MCM5
Genomic Map