siRNA Details
| Gene Symbol: Rpl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018860.4 | NP_061348.1 | 4 | 326 | ||
| GenBank mRNA | AK002503.1 | - | 4 | 272 | ||
| AK007874.1 | - | 4 | 274 | |||
| AK009548.1 | - | 4 | 273 | |||
| AK010246.1 | - | 4 | 273 | |||
| AK010317.1 | - | 4 | 273 | |||
| AK010482.1 | - | 4 | 273 | |||
| AK010509.1 | - | 4 | 273 | |||
| AK010564.1 | - | 4 | 271 | |||
| AK010583.1 | - | 4 | 273 | |||
| AK010585.1 | - | 4 | 271 | |||
| AK012252.1 | - | 4 | 271 | |||
| AK012450.1 | - | 4 | 276 | |||
| AK012476.1 | - | 4 | 272 | |||
| AK012487.1 | - | 4 | 271 | |||
| AK013324.1 | - | 4 | 278 | |||
| AK019049.1 | - | 4 | 273 | |||
| AK019178.1 | - | 4 | 268 | |||
| AK028126.1 | - | 4 | 271 | |||
| AK028231.1 | - | 4 | 274 | |||
| AK028263.1 | - | 4 | 271 | |||
| AK150502.1 | - | 4 | 273 | |||
| AK151566.1 | - | 4 | 272 | |||
| AK160588.1 | - | 4 | 273 | |||
| AK168107.1 | - | 4 | 273 | |||
| AK168485.1 | - | 4 | 271 | |||
| AK168653.1 | - | 4 | 271 | |||
| AK168719.1 | - | 4 | 271 | |||
| AK198849.1 | - | 1 | 41 | |||
| AV478133.1 | - | 4 | 259 | |||
| BC081450.1 | - | 3 | 342 | |||
| BC083066.1 | - | 4 | 260 | |||
| BC089572.1 | - | 4 | 270 | |||
| BC089573.1 | - | 4 | 272 | |||
| BC092010.1 | - | 4 | 261 | |||
| BC099380.1 | - | 3 | 340 | |||
| BC100592.1 | - | 4 | 272 | |||
| BC145727.1 | - | 4 | 273 | 40131030 | ||
| BC145729.1 | - | 4 | 273 | 40131032 | ||
| BU937034.1 | - | 4 | 329 | |||

Rpl41
Genomic Map