siRNA Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026470.3 | NP_080746.3 | 3, 4 | 436 | ||
| GenBank mRNA | AK051574.1 | - | 3, 4 | 437 | ||
| AK053170.1 | - | 3, 4 | 410 | |||
| AK132771.1 | - | 3, 4 | 410 | |||
| AK153098.1 | - | 3, 4 | 313 | |||

Spata6
Genomic Map