siRNA Details
| Gene Symbol: Sugt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026474.5 | NP_080750.1 | 9 | 597 | ||
| GenBank mRNA | AK007881.1 | - | 9 | 532 | ||
| AK010711.1 | - | 8 | 566 | |||
| AK010830.1 | - | 9 | 514 | |||
| AK020433.1 | - | 9 | 516 | |||
| AK045153.1 | - | 9 | 536 | |||
| AK082328.1 | - | 9 | 599 | |||
| AK141416.1 | - | 533 | ||||
| AK148838.1 | - | 9 | 511 | |||
| BC009167.1 | - | 9 | 514 | 2811924 | ||

Sugt1
Genomic Map