siRNA Details
| Gene Symbol: U2surp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114977.1 | NP_001108449.1 | 19 | 2033 | ||
| NM_026476.2 | NP_080752.2 | 18 | 1901 | |||
| GenBank mRNA | AK029898.1 | - | 19 | 1933 | ||
| AK161691.1 | - | 2 | 2575 | |||
| BC031497.1 | - | 19 | 1893 | |||
| BC068265.1 | - | 18 | 1917 | 30022109 | ||
| BC080699.1 | - | 18 | 1782 | |||

U2surp
Genomic Map