siRNA Details
| Gene Symbol: Ccny | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026484.3 | NP_080760.2 | 8 | 1080 | ||
| GenBank mRNA | AK017493.1 | - | 8 | 912 | ||
| AK028696.1 | - | 8 | 1081 | |||
| AK076556.1 | - | 8 | 610 | |||
| BC023321.1 | - | 9 | 998 | 3995040 | ||
| BC035524.1 | - | 7 | 835 | 4974858 | ||

Ccny
Genomic Map