siRNA Details
| Gene Symbol: Trabd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026485.2 | NP_080761.2 | 7 | 693 | ||
| GenBank mRNA | AK017745.1 | - | 7 | 687 | ||
| AK131116.1 | - | 1 | 2426 | |||
| AK146027.1 | - | 7 | 709 | |||
| AK185880.1 | - | 3 | 253 | |||
| BC005574.1 | - | 7 | 719 | 3967961 | ||
| BU510197.1 | - | 7 | 773 | |||

Trabd
Genomic Map