siRNA Details
| Gene Symbol: Trabd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026485.2 | NP_080761.2 | 4 | 378 | ||
| GenBank mRNA | AK005300.1 | - | 4 | 348 | ||
| AK017745.1 | - | 4 | 372 | |||
| AK146027.1 | - | 4 | 394 | |||
| BC005574.1 | - | 4 | 404 | 3967961 | ||
| BU510197.1 | - | 4 | 458 | |||
| CX200611.1 | - | 4 | 363 | |||

Trabd
Genomic Map