siRNA Details
| Gene Symbol: Atad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026487.3 | NP_080763.2 | 2 | 92 | ||
| GenBank mRNA | AK014967.1 | - | 2 | 109 | ||
| AK030719.1 | - | 2 | 112 | |||
| AK033639.1 | - | 2 | 116 | |||
| AK044925.1 | - | 2 | 92 | |||
| AK075685.1 | - | 2 | 136 | |||
| AK150469.1 | - | 2 | 112 | |||
| AK152059.1 | - | 2 | 112 | |||
| AK165953.1 | - | 2 | 127 | |||
| BC029085.1 | - | 2 | 92 | 3593451 | ||
| BC043051.1 | - | 2 | 77 | 5708377 | ||

Atad1
Genomic Map