siRNA Details
| Gene Symbol: Nacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037098.1 | NP_001032175.1 | 5 | 1910 | ||
| NM_026495.3 | NP_080771.3 | 5 | 1387 | |||
| GenBank mRNA | AK002651.1 | - | 5 | 1361 | ||
| AK147293.1 | - | 5 | 1387 | |||
| AK147310.1 | - | 5 | 1491 | |||
| AK157946.1 | - | 5 | 1494 | |||
| AK187605.1 | - | 2 | 82 | |||
| AK190138.1 | - | 2 | 139 | |||
| BC022103.1 | - | 5 | 1369 | 5067063 | ||
| BC070453.1 | - | 5 | 1690 | |||

Nacc2
Genomic Map