siRNA Details
| Gene Symbol: Nacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037098.1 | NP_001032175.1 | 6 | 2092 | ||
| NM_026495.3 | NP_080771.3 | 6 | 1569 | |||
| GenBank mRNA | AK002651.1 | - | 6 | 1543 | ||
| AK147293.1 | - | 6 | 1569 | |||
| AK147310.1 | - | 6 | 1673 | |||
| AK157946.1 | - | 6 | 1676 | |||
| BC022103.1 | - | 6 | 1551 | 5067063 | ||
| BC070453.1 | - | 6 | 1872 | |||

Nacc2
Genomic Map