siRNA Details
| Gene Symbol: Nacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037098.1 | NP_001032175.1 | 5, 6 | 1955 | ||
| NM_026495.3 | NP_080771.3 | 5, 6 | 1432 | |||
| GenBank mRNA | AK002651.1 | - | 5, 6 | 1406 | ||
| AK147293.1 | - | 5, 6 | 1432 | |||
| AK147310.1 | - | 5, 6 | 1536 | |||
| AK157946.1 | - | 5, 6 | 1539 | |||
| AK190138.1 | - | 2, 3 | 184 | |||
| BC022103.1 | - | 5, 6 | 1414 | 5067063 | ||
| BC070453.1 | - | 5, 6 | 1735 | |||

Nacc2
Genomic Map