siRNA Details
| Gene Symbol: Nudt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026497.2 | NP_080773.1 | 3 | 364 | ||
| GenBank mRNA | AK002641.1 | - | 3 | 365 | ||
| AK083846.1 | - | 3 | 322 | |||
| AK147867.1 | - | 3 | 318 | |||
| BC057657.1 | - | 3 | 318 | 2646355 | ||

Nudt12
Genomic Map