siRNA Details
| Gene Symbol: Nudt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026497.2 | NP_080773.1 | 6 | 1371 | ||
| GenBank mRNA | AK002641.1 | - | 6 | 1372 | ||
| AK083846.1 | - | 6 | 1329 | |||
| AK147867.1 | - | 6 | 1325 | |||
| BC050252.1 | - | 1 | 360 | |||

Nudt12
Genomic Map