siRNA Details
| Gene Symbol: Srsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026499.4 | NP_080775.3 | 4 | 648 | ||
| GenBank mRNA | AK004831.1 | - | 4 | 629 | ||
| AK012605.1 | - | 4 | 396 | |||
| AK051608.1 | - | 2 | 801 | |||
| AK133872.1 | - | 4 | 629 | |||
| AK146293.1 | - | 4 | 626 | |||
| AK159519.1 | - | 4 | 626 | |||
| AK171709.1 | - | 3 | 1912 | |||
| AK179065.1 | - | 3 | 277 | |||
| AK179529.1 | - | 2 | 234 | |||
| AK185344.1 | - | 1 | 96 | |||
| AK208133.1 | - | 1 | 95 | |||
| BC012039.1 | - | 4 | 635 | 2648111 | ||

Srsf6
Genomic Map