siRNA Details
| Gene Symbol: 1110004E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026502.2 | NP_080778.2 | 4 | 580 | ||
| GenBank mRNA | AK003413.1 | - | 4 | 471 | ||
| AK018510.1 | - | 4 | 551 | |||
| AK045905.1 | - | 4 | 559 | |||
| BC019533.1 | - | 4 | 544 | 4191729 | ||

1110004E09Rik
Genomic Map