siRNA Details
| Gene Symbol: Bambi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026505.2 | NP_080781.1 | 3 | 656 | ||
| GenBank mRNA | AF153440.1 | - | 3 | 642 | ||
| AK011305.1 | - | 3 | 679 | |||
| AK045192.1 | - | 3 | 656 | |||
| AK143185.1 | - | 3 | 671 | |||
| AK164632.1 | - | 3 | 659 | |||
| BC019378.1 | - | 3 | 638 | 5009872 | ||

Bambi
Genomic Map