siRNA Details
| Gene Symbol: SMCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030663.2 | NP_109588.2 | 2 | 740 | ||
| GenBank mRNA | BC014593.2 | - | 2 | 740 | 4824172 | |
| BC016744.2 | - | 2 | 723 | 4829283 | ||
| X89960.1 | - | 2 | 720 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMCP| Gene Symbol: SMCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030663.2 | NP_109588.2 | 2 | 740 | ||
| GenBank mRNA | BC014593.2 | - | 2 | 740 | 4824172 | |
| BC016744.2 | - | 2 | 723 | 4829283 | ||
| X89960.1 | - | 2 | 720 | |||