siRNA Details
| Gene Symbol: Col4a3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164222.1 | NP_001157694.1 | 11 | 1589 | ||
| NM_023420.2 | NP_075909.1 | 12 | 1667 | |||
| GenBank mRNA | AF232932.1 | - | 12 | 1681 | ||
| AF232934.1 | - | 11 | 1603 | |||
| AK012989.1 | - | 11 | 1570 | |||
| AK030305.1 | - | 12 | 1497 | |||
| AK080958.1 | - | 12 | 1665 | |||

Col4a3bp
Genomic Map