siRNA Details
| Gene Symbol: Col4a3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164222.1 | NP_001157694.1 | 13 | 1821 | ||
| NM_023420.2 | NP_075909.1 | 14 | 1899 | |||
| GenBank mRNA | AF232932.1 | - | 14 | 1913 | ||
| AF232934.1 | - | 13 | 1835 | |||
| AK012989.1 | - | 13 | 1802 | |||
| AK018103.1 | - | 2 | 190 | |||
| AK030305.1 | - | 14 | 1729 | |||

Col4a3bp
Genomic Map