siRNA Details
| Gene Symbol: Apopt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163388.1 | NP_001156860.1 | 3 | 371 | ||
| NM_026511.2 | NP_080787.1 | 3 | 371 | |||
| GenBank mRNA | AK002873.1 | - | 3 | 373 | ||
| AK005403.1 | - | 3 | 359 | |||
| AK010493.1 | - | 3 | 361 | |||
| AK012648.1 | - | 3 | 369 | |||
| AK029408.1 | - | 3 | 347 | |||
| AK148446.1 | - | 3 | 311 | |||
| AK160910.1 | - | 3 | 355 | |||
| BC071262.1 | - | 3 | 371 | 6509633 | ||

Apopt1
Genomic Map