siRNA Details
| Gene Symbol: ADAM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318933.1 | NP_001305862.1 | 9 | 750 | ||
| NM_002390.5 | NP_002381.2 | 9 | 754 | |||
| XM_005257373.3 | XP_005257430.1 | 9 | 896 | |||
| XM_017024647.1 | XP_016880136.1 | 8 | 668 | |||
| GenBank mRNA | AB009675.1 | - | 9 | 733 | ||
| AK296512.1 | - | 9 | 831 | |||
| D17390.1 | - | 6 | 463 | |||

ADAM11
Genomic Map