siRNA Details
| Gene Symbol: Cox19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197980.1 | NP_932097.1 | 2 | 150 | ||
| GenBank mRNA | AK002486.1 | - | 2 | 166 | ||
| AK003697.1 | - | 2 | 194 | |||
| AK007984.1 | - | 2 | 158 | |||
| AK013260.1 | - | 2 | 166 | |||
| AK013289.1 | - | 2 | 194 | |||
| AK148526.1 | - | 2 | 123 | |||
| BC031792.1 | - | 2 | 150 | 4193998 | ||

Cox19
Genomic Map