siRNA Details
| Gene Symbol: Rbm42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133693.2 | NP_598454.2 | 2 | 365 | ||
| GenBank mRNA | AK013929.1 | - | 2 | 303 | ||
| AK151000.1 | - | 2 | 277 | |||
| AK213653.1 | - | 1 | 18 | |||
| AK216265.1 | - | 1 | 18 | |||
| BC009148.1 | - | 2 | 302 | 3600406 | ||
| BC011286.1 | - | 2 | 284 | 3485410 | ||
| BC027372.1 | - | 2 | 289 | 4952304 | ||
| BC039917.1 | - | 2 | 300 | |||
| BC057928.1 | - | 2 | 291 | 5289543 | ||

Rbm42
Genomic Map