siRNA Details
| Gene Symbol: Zfp593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024215.2 | NP_077177.2 | 3 | 368 | ||
| GenBank mRNA | AK005247.1 | - | 3 | 370 | ||
| AK014081.1 | - | 2 | 412 | |||
| AK133585.1 | - | 3 | 366 | |||
| AK156931.1 | - | 3 | 303 | |||
| BC003465.1 | - | 3 | 305 | 2648747 | ||

Zfp593
Genomic Map