siRNA Details
| Gene Symbol: Chac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290667.1 | NP_001277596.1 | 1 | 180 | ||
| NM_026527.3 | NP_080803.1 | 1 | 180 | |||
| GenBank mRNA | AK007629.1 | - | 1 | 104 | ||
| AK010914.1 | - | 1 | 150 | |||
| AK021391.1 | - | 1 | 150 | |||
| AK078993.1 | - | 1 | 166 | |||
| BC025100.1 | - | 1 | 94 | 3967723 | ||
| BY333527.1 | - | 1 | 180 | |||

Chac2
Genomic Map