siRNA Details
| Gene Symbol: 2700062C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026529.4 | NP_080805.1 | 3 | 297 | ||
| GenBank mRNA | AK011228.1 | - | 2 | 384 | ||
| AK012467.1 | - | 2 | 399 | |||
| AK018391.1 | - | 3 | 301 | |||
| AK164298.1 | - | 1 | 2000 | |||
| BC029246.1 | - | 3 | 302 | |||
| BC037075.1 | - | 3 | 345 | 5038294 | ||
| BC084681.1 | - | 3 | 307 | 30144888 | ||

2700062C07Rik
Genomic Map