siRNA Details
| Gene Symbol: Chd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026539.2 | NP_080815.1 | 18 | 2173 | ||
| GenBank mRNA | AK014473.1 | - | 18 | 2175 | ||
| AK165656.1 | - | 18 | 2200 | |||
| BC020486.1 | - | 3 | 345 | |||
| BC052385.1 | - | 18 | 2157 | 5703859 | ||
| BC057567.1 | - | 18 | 2186 | 6415898 | ||
| BC062966.1 | - | 18 | 2186 | 6415898 | ||

Chd1l
Genomic Map