siRNA Details
| Gene Symbol: Tm9sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080556.3 | NP_542123.3 | 15 | 1886 | ||
| GenBank mRNA | AK030340.1 | - | 15 | 1817 | ||
| AK048012.1 | - | 15 | 1805 | |||
| AK050338.1 | - | 15 | 1833 | |||
| AK075720.1 | - | 15 | 1886 | |||
| AK088905.1 | - | 15 | 1858 | |||
| AK159757.1 | - | 15 | 1831 | |||
| BC003862.1 | - | 15 | 1825 | 3488326 | ||

Tm9sf2
Genomic Map