siRNA Details
| Gene Symbol: Gpn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024216.1 | NP_077178.1 | 6 | 719 | ||
| GenBank mRNA | AK016996.1 | - | 6 | 650 | ||
| AK042405.1 | - | 1434 | ||||
| AK080720.1 | - | 6 | 666 | |||
| AK087968.1 | - | 6 | 670 | |||
| AK146783.1 | - | 6 | 657 | |||
| BC003341.1 | - | 6 | 719 | 3257412 | ||

Gpn3
Genomic Map