siRNA Details
| Gene Symbol: Dcakd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026551.3 | NP_080827.2 | 3, 4 | 514 | ||
| GenBank mRNA | AK051022.1 | - | 3, 4 | 515 | ||
| AK078460.1 | - | 3, 4 | 447 | |||
| AK162069.1 | - | 3, 4 | 474 | |||
| AK170762.1 | - | 3, 4 | 465 | |||
| BC005607.1 | - | 3, 4 | 480 | 3498151 | ||
| BC023208.1 | - | 2, 3 | 428 | |||
| BC031855.1 | - | 3, 4 | 463 | |||
| BC042685.1 | - | 2, 3 | 428 | |||
| BC045193.1 | - | 3, 4 | 468 | 5368157 | ||
| BC047340.1 | - | 2, 3 | 428 | |||

Dcakd
Genomic Map