siRNA Details
| Gene Symbol: Yif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026553.4 | NP_080829.1 | 6 | 803 | ||
| GenBank mRNA | AK002259.1 | - | 2 | 208 | ||
| AK011071.1 | - | 6 | 798 | |||
| AK077329.1 | - | 6 | 789 | |||
| AK160409.1 | - | 6 | 756 | |||
| BC011117.1 | - | 6 | 745 | 4190535 | ||

Yif1a
Genomic Map