siRNA Details
| Gene Symbol: Sdccag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085407.1 | NP_001078876.1 | 6, 7 | 940 | ||
| NM_001085408.1 | NP_001078877.1 | 5, 6 | 871 | |||
| NM_026563.3 | NP_080839.2 | 7, 8 | 1090 | |||
| GenBank mRNA | AK014885.1 | - | 7, 8 | 1059 | ||
| AK016227.1 | - | 1, 2 | 34 | |||
| AK082787.1 | - | 6, 7 | 911 | |||
| AK087227.1 | - | 1, 2 | 456 | |||
| AK163690.1 | - | 5, 6 | 842 | |||
| BC031199.1 | - | 5, 6 | 820 | 5064948 | ||

Sdccag3
Genomic Map