siRNA Details
| Gene Symbol: Sdccag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085407.1 | NP_001078876.1 | 4 | 491 | ||
| NM_001085408.1 | NP_001078877.1 | 3 | 422 | |||
| NM_026563.3 | NP_080839.2 | 5 | 641 | |||
| GenBank mRNA | AK021273.1 | - | 5 | 632 | ||
| AK082787.1 | - | 4 | 462 | |||
| AK163690.1 | - | 3 | 393 | |||
| AW743523.1 | - | 3 | 372 | |||
| BC005612.1 | - | 3 | 413 | |||
| BC031199.1 | - | 3 | 371 | 5064948 | ||
| BC061037.1 | - | 3 | 702 | |||

Sdccag3
Genomic Map