siRNA Details
| Gene Symbol: 9430016H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081181.2 | NP_001074650.1 | 2 | 714 | ||
| GenBank mRNA | AK007831.1 | - | 2 | 503 | ||
| AK079116.1 | - | 2 | 716 | |||
| AK165130.1 | - | 1 | 3198 | |||
| BC071241.1 | - | 511 | 5125554 | |||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Rat: NM_001004251, BC079246.
					            	This siRNA also targets the following transcripts in Rat: NM_001004251, BC079246.Gene Ontology Categories:
 Back To Top
 Back To Top
 9430016H08Rik
9430016H08Rik Genomic Map
Genomic Map Back To Top
 Back To Top