siRNA Details
| Gene Symbol: Cep70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023873.3 | NP_076362.2 | 3, 4 | 257 | ||
| GenBank mRNA | AF242319.1 | - | 3, 4 | 293 | ||
| AK020174.1 | - | 3, 4 | 595 | |||
| AK020622.1 | - | 3, 4 | 334 | |||
| AK021095.1 | - | 3, 4 | 388 | |||
| AK049326.1 | - | 3, 4 | 265 | |||
| AK133048.1 | - | 3, 4 | 263 | |||
| BC050819.1 | - | 3, 4 | 249 | 6517145 | ||
| BC071260.1 | - | 3, 4 | 257 | 6336351 | ||

Cep70
Genomic Map